2-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049802-500MG 049802-2.5G
101889-108EA 364.62 USD
101889-108 101889-110
2-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride
2-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClFNO
MDL Number: MFCD13560204
CAS Number: 1219957-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR