3-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride

Supplier: Matrix Scientific

049801-500MG 049801-2.5G
101889-104EA 364.62 USD
101889-104 101889-106
3-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride
3-[2-(2-Fluoroethoxy)ethyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₉H₁₉ClFNO
MDL Number: MFCD13560203
CAS Number: 1220016-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR