3-Fluoro-5-hydroxybenzoic acid ≥98%

Supplier: Matrix Scientific

059598-1G 059598-5G
101918-610EA 20 USD
101918-610 101918-612
3-Fluoro-5-hydroxybenzoic acid ≥98%
3-Fluoro-5-hydroxybenzoic acid

Hazard: Irritant

Formula: C₇H₅FO₃
MDL Number: MFCD11846515
CAS Number: 860296-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR