3-[(4-Iodobenzyl)oxy]piperidine hydrochloride

Supplier: Matrix Scientific

050838-500MG 050838-2.5G
101893-248EA 364.62 USD
101893-248 101893-250
3-[(4-Iodobenzyl)oxy]piperidine hydrochloride
3-[(4-Iodobenzyl)oxy]piperidine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClINO
MDL Number: MFCD13561218
CAS Number: 1219977-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR