2-Fluoro-5-(trifluoromethyl)benzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanone

002626-1G 002626-5G
101784-330EA 30.77 USD
101784-330 101784-332
2-Fluoro-5-(trifluoromethyl)benzophenone ≥97%
2-Fluoro-5-(trifluoromethyl)benzophenone

Hazard: Irritant

Formula: C₁₄H₈F₄O
MDL Number: MFCD00061254
CAS Number: 199292-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR