2-Fluoro-5-iodotoluene ≥99%

Supplier: Matrix Scientific
Warning

002706-5G 002706-25G
101784-484EA 29.23 USD
101784-484 101784-486
2-Fluoro-5-iodotoluene ≥99%
2-Fluoro-5-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆FI
Boiling Pt: 86…87 °C (9mm)
MDL Number: MFCD00013710
CAS Number: 452-68-6
EINECS: 207-206-1

Order Now

Specification Test Results

Boiling point 86-87°C/9mm
ND20 1.578

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR