Sodium chromate tetrahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-1129-500G 93-1129-100G
100204-666EA 188.25 USD
100204-666 100197-668
Sodium chromate tetrahydrate ≥99%
Sodium chromate tetrahydrate
Formula: CrNa₂O₄·4H₂O
Melting Pt: 792.2 °C
Density: 2.73 g/cm³ (20 °C)
MDL Number: MFCD00149167
CAS Number: 10034-82-9
EINECS: 231-889-5
UN: 3087
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR