D-(-)-Tartaric acid ≥99%

Warning

T206-100G T206-25G
220226-150EA 131.38 USD
220226-150 220226-151
D-(-)-Tartaric acid ≥99%
D-(-)-Tartaric acid
Formula: C₄H₆O₆
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 166…169 °C
Density: 1.76 g/cm³ (20 °C)
Flash Pt: 210 °C
Storage Temperature: Ambient
MDL Number: MFCD00004238
CAS Number: 147-71-7
EINECS: 205-695-6
Merck Index: 12,09235

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR