D-(-)-Sorbitol ≥98%

S1876-10MG S1876-1KG S1876-10KG S1876-500G S1876-100G S1876-5KG
220208-638EA 32 USD
220208-638 220208-639 220208-637 220208-640 220208-636 220208-641
D-(-)-Sorbitol ≥98%
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 295 °C (1013 hPa)
Melting Pt: 94…98 °C
Density: 1.524 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
Merck Index: 13,08797
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR