Barium chromate ≥98%

Danger

401056-250G 401056-1KG
220061-952EA 127.69 USD
220061-952 220061-951
Barium chromate ≥98%
Barium chromate
Formula: BaCrO₄
Melting Pt: 1400 °C
Density: 4.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014180
CAS Number: 10294-40-3
EINECS: 233-660-5
UN: 1479
ADR: 5.1,III
Merck Index: 12,00999

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR