D-(-)-Sorbitol ≥70% (w/w) in aqueous solution

309532-500ML 309532-100ML
220054-461EA 178.46 USD
220054-461 220054-460
D-(-)-Sorbitol ≥70% (w/w) in aqueous solution
D(-)-Sorbitol
Formula: HOH₂C(CH(OH))₄CH₂OH
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004708
CAS Number: 50-70-4
EINECS: 200-061-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82054e8cfed2cf3e8b739c3cb816c4f91bd2735&vid=a084778ac67fbdf7e15478c0db755e78cf3d95c9&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR