2,6-Di-tert-butylpyridine

Warning

219584-5G 219584-25G 219584-1G
220044-668EA 199.23 USD
220044-668 220044-667 220044-666
2,6-Di-tert-butylpyridine
2,6-Di-tert-butylpyridine
Formula: C₁₃H₂₁N
Boiling Pt: 209 °C (1013 hPa)
Density: 0.852 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00006306
CAS Number: 585-48-8
EINECS: 209-557-6
Merck Index: 12,03086

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR