Sodium chromate tetrahydrate ≥99%

Danger

216623-100G 216623-500G
220044-422EA 38.08 USD
220044-422 220044-423
Sodium chromate tetrahydrate ≥99%
Sodium chromate tetrahydrate
Formula: CrNa₂O₄·4H₂O
Melting Pt: 792.2 °C
Density: 2.73 g/cm³ (20 °C)
MDL Number: MFCD00149167
CAS Number: 10034-82-9
EINECS: 231-889-5
UN: 3288
ADR: 6.1,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR