Ammonium cerium(IV) nitrate

Danger

229547-250G 229547-10G 229547-50G
220045-841EA 1138.46 USD
220045-841 220045-840 220045-842
Ammonium cerium(IV) nitrate
Ammonium cerium (IV) nitrate
Formula: (NH₄)₂Ce(NO₃)₆
Melting Pt: 107…108 °C
Density: 2.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151121
CAS Number: 16774-21-3
EINECS: 240-827-6
UN: 1477
ADR: 5.1,III
Merck Index: 13,00509

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR