Horizontal Laminar Flow Clean Benches, Envirco®
Supplier: Envirco
10555 10552 10553
VF-10555EA 36033.01 USD
VF-10555 VF-10552 VF-10553
Horizontal Laminar Flow Clean Benches, Envirco®
Fume Hoods and Enclosures Laminar Flow Cabinets
The two-piece design eliminates nearly all vibration transmission from filter/blower console to work surface
The workstation frame is made of welded steel with a corrosion-resistant polyurethane enamel finish. The work surface has a depth of 61cm (24") and is 76.2Hcm (30") from floor. Benches feature removable acrylic side panels and a convenient recessed knee space for operator comfort. A duplex receptacle is located on the front leg of the bench for easy access. The nonglare, plastic laminated work surface is easy to clean. A recessed fluorescent light fixture provides 150 foot-candles of illumination at the work surface.
Benches utilize a direct-drive continuous-duty 1/2hp motor with sealed-for-life bearings dynamically balanced and mounted on rubber-in-shear vibration isolators. The motor is designed to provide rated airflow through the filter loading in excess of 50% over initial static pressure. The motor features solid-state speed control with RFI suppression for adjusting airflow velocity.
Benches feature HEPA filters to provide an ISO Class 5 (FED-STD-209E Class 100/M3.5) work environment. The HEPA filters have a minimum efficiency of 99.99% at 0.3µm or larger. A perforated aluminum diffuser screen protects the HEPA filter while maintaining uniform air velocity within the work area. A 30% ASHRAE washable prefilter is included to extend the life of the HEPA filter.
Información para pedidos: Manufacturer's one-year limited warranty on parts and labor. Include a three-wire cord with plug. Accessory items such as stainless steel work surfaces, lab service fixtures, and IV rods are available; contact your Avantor sales representative for more information.