5-Aminoindan ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A151769-1G A151769-25G A151769-5G A151769-100G
ALA151769-1GEA 11.31 USD
ALA151769-1G ALA151769-25G ALA151769-5G ALA151769-100G
5-Aminoindan ≥98%
5-Aminoindan

Product Application:5-Aminoindan was used in the synthesis of ligand 18/18 (2-(5-aminoindan)-6-(5-aminoindan)-4-chloro-s-triazine).

Formula: C₉H₁₁N
Boiling Pt: 147°C
Melting Pt: 34-38°C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003803
CAS Number: 24425-40-9
EINECS: 246-241-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eaebfee469d773fd42fab10df35d75b07023ab34&vid=eaebfee469d773fd42fab10df35d75b07023ab34&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR