1,5-Anhydro-D-sorbitol ≥97%

Supplier: ALADDIN SCIENTIFIC CORPORATION
Warning

A121074-100MG A121074-500MG
ALA121074100MGEA 583.9 USD
ALA121074100MG ALA121074500MG
1,5-Anhydro-D-sorbitol ≥97%
1,5-Anhydro-D-sorbitol

1,5-Anhydro-D-sorbitol, a cyclic polyol, is a metabolic product of 1,5 anhydro-D-fructose.1,5-Anhydro-D-sorbitol is used as a reference in analytical procedures that assess the metabolism of 1,5 anhydro-D-fructose, a food supplement with potential benefits as an antioxidant, antimicrobial, and antidiabetic agent.

Formula: C₆H₁₂O₅
MDL Number: MFCD00067386
CAS Number: 154-58-5
EINECS: 205-829-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fc033b7eeee0a0473b3354ca961b25bd13b4f2e5&vid=fc033b7eeee0a0473b3354ca961b25bd13b4f2e5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR