Ethyl 2-(3,5-di-tert-butylphenyl)-2-oxoacetate ≥95%

Supplier: AOBChem USA

Warning

82608-5G 82608-1G 82608-500MG
77630-704EA 1537 USD
77630-704 77644-074 77648-040
Ethyl 2-(3,5-di-tert-butylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(3,5-di-tert-butylphenyl)-2-oxoacetate

Precaución: For Research Use Only

Formula: 290.397 g/mol
MW: C₁₈H₂₆O₃
Storage Temperature: Ambient
MDL Number: MFCD11555596
CAS Number: 1256466-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR