α-Ethenyl-4-fluoro-2-methoxybenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

355988-5G 355988-1G 355988-500MG
77628-774EA 1484.23 USD
77628-774 77638-306 77650-328
α-Ethenyl-4-fluoro-2-methoxybenzenemethanol ≥95%
α-Ethenyl-4-fluoro-2-methoxybenzenemethanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD35189943
CAS Number: 2228606-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR