1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

325562-5G 325562-25G 325562-1G 325562-500MG
77628-606EA 1484.23 USD
77628-606 77655-062 77642-072 77646-072
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)propan-1-one ≥95%
1-(2,3-Difluoro-4-(trifluoromethyl)phenyl)propan-1-one

Precaución: For Research Use Only

Formula: C₁₀H₇F₅O
Storage Temperature: Ambient
MDL Number: MFCD35178668
CAS Number: 3002541-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR