Methanone, (4-chloro-2-fluoro-3-methylphenyl)phenyl- ≥95%

Supplier: AOBChem USA

Warning

250483-5G 250483-1G 250483-500MG
77628-232EA 1484.23 USD
77628-232 77637-466 77645-456
Methanone, (4-chloro-2-fluoro-3-methylphenyl)phenyl- ≥95%
Methanone, (4-chloro-2-fluoro-3-methylphenyl)phenyl-

Precaución: For Research Use Only

Formula: C₁₄H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD34873066
CAS Number: 1502528-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR