S116836 95%

Supplier: Ambeed
Warning

A1532005-25MG A1532005-100MG A1532005-250MG A1532005-50MG
77625-278EA 330.63 USD
77625-278 77876-135 77876-136 77876-137
S116836 95%
S116836

Precaución: for Research Use Only

Formula: C₂₇H₂₁F₃N₆O
Storage Temperature: Refrigerator
CAS Number: 1257628-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR