2-Methoxy-4-pyridineboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Methoxy-4-pyridylboronic acid, 2-Methoxypyridine-4-boronic acid, (2-Methoxypyridin-4-yl)boronic acid
M2730-1G
M2730-5G
TCM2730-1GEA
123
USD
TCM2730-1G
TCM2730-5G
2-Methoxy-4-pyridineboronic acid (contains varying amounts of Anhydride)
2-Methoxy-4-pyridineboronic acid
Formula:
C₆H₈BNO₃ |
MDL Number:
MFCD07368877 CAS Number: 762262-09-9 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | 97.0 to 114.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...