InVivoMAb Anti-E protein DI-DII Mouse Monoclonal Antibody [clone: E53]

Supplier: Bio X Cell
BE0430-100MG BE0430-1MG BE0430-25MG BE0430-50MG BE0430-5MG
77579-840EA 4408.4 USD
77579-840 77579-842 77579-844 77579-846 77579-848
InVivoMAb Anti-E protein DI-DII Mouse Monoclonal Antibody [clone: E53]
Antibodies
Type: Primary
Antigen: E protein DI-DII
Clonality: Monoclonal
Clone: E53
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: West nile virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR