InVivoPlus Anti-CD71 Rat Antibody [clone: R17]

Supplier: Bio X Cell
BP0175-50MG BP0175-5MG BP0175-100MG BP0175-25MG
77579-980EA 4054.08 USD
77579-980 77579-982 77579-976 77579-978
InVivoPlus Anti-CD71 Rat Antibody [clone: R17]
Antibodies
Type: Primary
Antigen: CD71
Clonality:
Clone: R17
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR