Di-tert-butyl 3,3'-((2-amino-2-((3-(tert-butoxy)-3-oxopropoxy)methyl)propane-1,3-diyl)bis(oxy))dipropanoate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Tris[[2-(tert-butoxycarbonyl)ethoxy]methyl]methylamine

A904817-250MG A904817-25G A904817-5G A904817-100MG A904817-1G
77684-862EA 19 USD
77684-862 77891-674 77891-675 77891-672 77891-673
Di-tert-butyl 3,3'-((2-amino-2-((3-(tert-butoxy)-3-oxopropoxy)methyl)propane-1,3-diyl)bis(oxy))dipropanoate ≥95%, Ambeed.Inc
Di-tert-butyl 3,3'-((2-amino-2-((3-(tert-butoxy)-3-oxopropoxy)methyl)propane-1,3-diyl)bis(oxy))dipropanoate

Precaución: For research use only.

Formula: C₂₅H₄₇NO₉
MDL Number: MFCD18252299
CAS Number: 175724-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR