U-73122 ≥98%

Supplier: Ambeed
Warning

A877124-1MG A877124-5MG A877124-10MG
77595-418EA 29.73 USD
77595-418 77674-308 77674-306
U-73122 ≥98%
U-73122

Precaución: for Research Use Only

Formula: C₂₉H₄₀N₂O₃
MDL Number: MFCD00893825
CAS Number: 112648-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR