6-Bromobenzo[d]thiazole-2-carbonitrile ≥95%

Supplier: Ambeed
Warning

A662978-1G A662978-100MG A662978-250MG
77673-810EA 292 USD
77673-810 77884-542 77884-543
6-Bromobenzo[d]thiazole-2-carbonitrile ≥95%
6-Bromobenzo[d]thiazole-2-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₃BrN₂S
MDL Number: MFCD11217379
CAS Number: 741253-03-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR