4-Bromo-5-fluoro-1H-indazole ≥97%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A612750-100MG
A612750-1G
A612750-5G
77684-489EA
32
USD
77684-489
77886-471
77886-472
4-Bromo-5-fluoro-1H-indazole ≥97%, Ambeed.Inc
4-Bromo-5-fluoro-1H-indazole
Precaución: For research use only.
Formula:
C₇H₄BrFN₂ |
MDL Number:
MFCD16987597 CAS Number: 1056264-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...