Molybdic acid 85.0% (MoO₃ basis) AR

Supplier: Ambeed
Warning

A555890-500G A555890-100G
77602-362EA 66.63 USD
77602-362 77669-464
Molybdic acid 85.0% (MoO₃ basis) AR
Molybdic acid

Precaución: for Research Use Only

Formula: H₂MoO₄
MDL Number: MFCD00036287
CAS Number: 7782-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR