4-Chloro-α,α-dimethylphenylacetic acid ≥98%

Supplier: Ambeed
Warning

A551954-250MG A551954-5G
77878-339EA 14 USD
77878-339 77878-340
4-Chloro-α,α-dimethylphenylacetic acid ≥98%
4-Chloro-alpha,alpha-dimethylphenylacetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MDL Number: MFCD00044814
CAS Number: 6258-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR