Chroman-8-ylboronic acid ≥95%

Supplier: Ambeed
Warning

A529331-100MG A529331-1G A529331-250MG
77891-071EA 38 USD
77891-071 77891-072 77891-073
Chroman-8-ylboronic acid ≥95%
Chroman-8-ylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₃
MDL Number: MFCD09266209
CAS Number: 685514-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR