Methyl-4-amino-3-chloro-5-nitrobenzoate ≥98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-3-chloro-5-nitrobenzoate

A504863-1G A504863-5G
77613-588EA 9.27 USD
77613-588 77891-025
Methyl-4-amino-3-chloro-5-nitrobenzoate ≥98%
Methyl 4-amino-3-chloro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₄
MDL Number: MFCD09842615
CAS Number: 863886-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR