3-Amino-2-hydroxybenzotrifluoride ≥96%

Supplier: Ambeed
Warning

A345799-100MG A345799-250MG
77885-910EA 29 USD
77885-910 77885-911
3-Amino-2-hydroxybenzotrifluoride ≥96%
3-Amino-2-hydroxybenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₆F₃NO
MDL Number: MFCD08532490
CAS Number: 72534-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR