2-Chloro-5-nitroquinoline ≥98%

Supplier: Ambeed
Warning

A192822-1G A192822-250MG A192822-25G A192822-5G
77885-429EA 32 USD
77885-429 77885-430 77885-431 77885-432
2-Chloro-5-nitroquinoline ≥98%
2-Chloro-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MDL Number: MFCD18448913
CAS Number: 13067-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR