(3-Methylisoxazol-5-yl)methylamine ≥98%

Supplier: Ambeed
Danger

A177141-5G A177141-100MG A177141-250MG
77671-514EA 794 USD
77671-514 77883-346 77883-347
(3-Methylisoxazol-5-yl)methylamine ≥98%
(3-Methylisoxazol-5-yl)methylamine

Precaución: for Research Use Only

Formula: C₅H₈N₂O
MDL Number: MFCD06738858
CAS Number: 154016-55-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR