tert-Butyl 2-(prop-2-yn-1-yloxy)acetate ≥98%

Supplier: Ambeed
Warning

A1726559-1G A1726559-100MG
77598-220EA 191.68 USD
77598-220 77877-191
tert-Butyl 2-(prop-2-yn-1-yloxy)acetate ≥98%
tert-Butyl 2-(prop-2-yn-1-yloxy)acetate

Precaución: for Research Use Only

Formula: C₉H₁₄O₃
MDL Number: MFCD29087247
CAS Number: 339531-55-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR