(R)-2-Phenylpyrrolidine hydrochloride ≥95%

Supplier: Ambeed
Warning

A154719-1G A154719-250MG
77890-009EA 105 USD
77890-009 77890-010
(R)-2-Phenylpyrrolidine hydrochloride ≥95%
(R)-2-Phenylpyrrolidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃N·ClH
MDL Number: MFCD12910596
CAS Number: 56523-48-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR