2-Chloro-5-hydroxy-3-methylpyridine ≥96%

Supplier: Ambeed
Warning

A117614-5G A117614-10G A117614-25G
77670-650EA 166 USD
77670-650 77882-962 77882-963
2-Chloro-5-hydroxy-3-methylpyridine ≥96%
2-Chloro-5-hydroxy-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MDL Number: MFCD11519295
CAS Number: 54232-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR