3-Methoxy-2,4,6-trifluorophenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (2,4,6-Trifluoro-3-methoxyphenyl)boronic acid

A103596-1G A103596-5G
77882-883EA 53 USD
77882-883 77882-884
3-Methoxy-2,4,6-trifluorophenylboronic acid ≥98%
3-Methoxy-2,4,6-trifluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₃
MDL Number: MFCD07369755
CAS Number: 849062-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR