But-3-yn-1-amine 98%

Supplier: Ambeed
Danger

A902473-1G A902473-250MG
77552-696EA 119 USD
77552-696 77569-632
But-3-yn-1-amine 98%
But-3-yn-1-amine

Precaución: For research use only.

Formula: C₄H₇N
MW: 69.11 g/mol
MDL Number: MFCD09997741
CAS Number: 14044-63-4
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR