1-(2,3-Difluoro-4-(methoxymethoxy)phenyl)-2,2-Difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

75573-1G 75573-500MG 75573-5G 75573-25G
77473-088EA 415.09 USD
77473-088 77485-706 77496-294 77505-402
1-(2,3-Difluoro-4-(methoxymethoxy)phenyl)-2,2-Difluoroethanone ≥95%
1-(2,3-Difluoro-4-(methoxymethoxy)phenyl)-2,2-Difluoroethanone

Precaución: For research use only

Formula: C₁₀H₈F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34766068
CAS Number: 2901065-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR