α-Cyclohexyl[1,1'-biphenyl]-4-methanol ≥95%

Supplier: AOBChem USA

Warning

186843-1G 186843-500MG 186843-5G 186843-25G
77466-324EA 415.09 USD
77466-324 77479-068 77489-304 77499-668
α-Cyclohexyl[1,1'-biphenyl]-4-methanol ≥95%
α-Cyclohexyl[1,1'-biphenyl]-4-methanol

Precaución: For research use only

Formula: C₁₉H₂₂O
Storage Temperature: Ambient
MDL Number: MFCD34859238
CAS Number: 1071934-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR