(2,3-Difluoro-4-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol ≥95%

Supplier: AOBChem USA

325568-1G 325568-500MG 325568-5G 325568-25G
77469-346EA 415.09 USD
77469-346 77482-022 77492-426 77502-314
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol ≥95%
(2,3-Difluoro-4-(trifluoromethyl)phenyl)(4-ethylphenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₃F₅O
CAS Number: 3002524-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR