2-Chloro-1-(2,3-difluoro-4-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

325564-1G 325564-500MG 325564-5G 325564-25G
77469-342EA 415.09 USD
77469-342 77482-018 77492-422 77502-310
2-Chloro-1-(2,3-difluoro-4-(trifluoromethyl)phenyl)-2,2-difluoroethanone ≥95%
2-Chloro-1-(2,3-difluoro-4-(trifluoromethyl)phenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₉H₂ClF₇O
CAS Number: 3002422-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR