4-Methyl-2-(pyridin-4-yl)-1,3-thiazole-5-carboxaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR14888-100MG OR14888-1G OR14888-250MG
77405-152EA 48.9 USD
77405-152 77405-154 77405-156
4-Methyl-2-(pyridin-4-yl)-1,3-thiazole-5-carboxaldehyde 97%
4-Methyl-2-(pyridin-4-yl)-1,3-thiazole-5-carboxaldehyde
Formula: C₁₀H₈N₂OS
Melting Pt: 168…170 °C
Storage Temperature: Ambient
MDL Number: MFCD09817496
CAS Number: 892502-19-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR