1,1,1,2,2-Pentafluoropentane

Supplier: APOLLO SCIENTIFIC
Danger

PC450571-1G PC450571-250MG PC450571-5G
77415-576EA 137.54 USD
77415-576 77415-578 77415-580
1,1,1,2,2-Pentafluoropentane
1,1,1,2,2-Pentafluoropentane
Formula: C₅H₇F₅
Storage Temperature: Ambient
MDL Number: MFCD28135687
CAS Number: 154381-43-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR