2-Bromo-4-cyclobutyl-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

49493-500MG 49493-1G 49493-5G
77336-730EA 393.46 USD
77336-730 77351-506 77375-780
2-Bromo-4-cyclobutyl-1-methoxybenzene ≥97%
2-Bromo-4-cyclobutyl-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34178569
CAS Number: 2643367-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR