2-Fluoro-5-propoxyphenol ≥97%

Supplier: AOBChem USA

Warning

46134-500MG 46134-1G 46134-5G 46134-25G
77327-060EA 201.88 USD
77327-060 77350-644 77363-362 77386-002
2-Fluoro-5-propoxyphenol ≥97%
2-Fluoro-5-propoxyphenol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD29044445
CAS Number: 1881290-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR