(4-Butoxy-2,3-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

37449-1G 37449-5G 37449-25G
77341-546EA 92.7 USD
77341-546 77362-690 77374-286
(4-Butoxy-2,3-difluorophenyl)(methyl)sulfane ≥97%
(4-Butoxy-2,3-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₁H₁₄F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34529933
CAS Number: 2734777-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR